missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium Fluoride Hydrate 98.0+%, TCI America™
Supplier: TCI America T1037100G
Specifications
| Tetrabutylammonium Fluoride Hydrate [for Catalyst of acylation, silylation and cleavage of silyl eth | |
| White | |
| MFCD00011747 | |
| 1759 | |
| UQCWXKSHRQJGPH-UHFFFAOYSA-N | |
| tetrabutylazanium;hydrate;hydrofluoride | |
| 123134460 | |
| ≥98.0% (T) |
| 22206-57-1 | |
| C16H39FNO+ | |
| 100 g | |
| TBAF | |
| CCCC[N+](CCCC)(CCCC)CCCC.O.F | |
| 280.492 | |
| 261.47 | |
| Crystalline Powder |
Chemical Identifiers
| 22206-57-1 | |
| 280.492 | |
| UQCWXKSHRQJGPH-UHFFFAOYSA-N | |
| 123134460 | |
| CCCC[N+](CCCC)(CCCC)CCCC.O.F |
| C16H39FNO+ | |
| MFCD00011747 | |
| TBAF | |
| tetrabutylazanium;hydrate;hydrofluoride |
Safety and Handling
EINECSNumber : (2)-0186
TSCA : Yes