missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Fluoride Hydrate 98.0+%, TCI America™

Catalog No. T103725G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T103725G 25 g
T1037100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T103725G Supplier TCI America Supplier No. T103725G
Only null left

Chemical Identifiers

CAS 22206-57-1
Molecular Formula C16H39FNO+
Molecular Weight (g/mol) 280.492
MDL Number MFCD00011747
InChI Key UQCWXKSHRQJGPH-UHFFFAOYSA-N
Synonym TBAF
PubChem CID 123134460
IUPAC Name tetrabutylazanium;hydrate;hydrofluoride
SMILES CCCC[N+](CCCC)(CCCC)CCCC.O.F

Specifications

Color White
Quantity 25 g
UN Number 1759
Formula Weight 261.47
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Fluoride Hydrate [for Catalyst of acylation, silylation and cleavage of silyl eth
EINECSNumber (2)-0186
TSCA Yes