missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Dihydrogen Trifluoride 90.0+%, TCI America™

Catalog No. T16355G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16355G 5 g
T163525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16355G Supplier TCI America Supplier No. T16355G

Chemical Identifiers

CAS 99337-56-1
Molecular Formula C16H38F3N
Molecular Weight (g/mol) 301.48
MDL Number MFCD00145365
InChI Key MRXQMNWIADOAJY-UHFFFAOYSA-M
Synonym tetra-n-butylammonium dihydrogen trifluoride, tetra-n-butylammonium dihydrogentrifluoride, dihydrogen tetrabutylammonium fluoride, tetrabutylammonium dihydrogentrifluoride, tetrabutylammonium dihydrogen trifluoride, tetrabutylammonium fluoride dihydrofluoride, tetrabutylazanium fluoride dihydrofluoride, tetrabutylammoniumdihydrogentrifluoride, tetrabutylamonium dihydrogentrifluoride, tetrabutylammoniumdihydrogen trifluoride
PubChem CID 11748636
IUPAC Name tetrabutylazanium dihydrofluoride fluoride
SMILES F.F.[F-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Color Yellow
Quantity 5 g
UN Number 1760
Formula Weight 301.48
Percent Purity ≥90.0% (T)
Physical Form Liquid
Chemical Name or Material Tetrabutylammonium Dihydrogen Trifluoride
EINECSNumber (2)-0186
TSCA No