missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium Difluorotriphenylstannate 97.0+%, TCI America™
Supplier: TCI America T15925G
Specifications
| Tetrabutylammonium Difluorotriphenylstannate | |
| 193°C | |
| C34H51F2NSn | |
| 5 g | |
| tetrabutylammonium difluorotriphenylstannate, tetrabutylammonium difluorotriphenylstannate iv, tetrabutylammonium difluorotriphenyltin, tetrabutylammonium triphenyldifluorotin, tetrabutylammoniumdifluorotriphenylstannate, tetrabutylam-monium triphenyldifluorostannate, tetrabutylammonium difluorotriphenyl stannate, tetra-n-butylammonium difluorotriphenylstannate, difluorotriphenylstannanuide; tetrabutylammonium, difluoro triphenyl stannanuide; tetrabutylammonium | |
| CCCC[N+](CCCC)(CCCC)CCCC.C1=CC=C(C=C1)[Sn-](C2=CC=CC=C2)(C3=CC=CC=C3)(F)F | |
| 630.496 | |
| 630.50 | |
| Crystalline Powder |
| 139353-88-1 | |
| White-Yellow | |
| MFCD00192465 | |
| 3146 | |
| ODMXVCNGZGLSRS-UHFFFAOYSA-L | |
| difluoro(triphenyl)stannanuide;tetrabutylazanium | |
| 2733194 | |
| ≥97.0% (T) |
Chemical Identifiers
| 139353-88-1 | |
| 630.496 | |
| ODMXVCNGZGLSRS-UHFFFAOYSA-L | |
| 2733194 | |
| CCCC[N+](CCCC)(CCCC)CCCC.C1=CC=C(C=C1)[Sn-](C2=CC=CC=C2)(C3=CC=CC=C3)(F)F |
| C34H51F2NSn | |
| MFCD00192465 | |
| tetrabutylammonium difluorotriphenylstannate, tetrabutylammonium difluorotriphenylstannate iv, tetrabutylammonium difluorotriphenyltin, tetrabutylammonium triphenyldifluorotin, tetrabutylammoniumdifluorotriphenylstannate, tetrabutylam-monium triphenyldifluorostannate, tetrabutylammonium difluorotriphenyl stannate, tetra-n-butylammonium difluorotriphenylstannate, difluorotriphenylstannanuide; tetrabutylammonium, difluoro triphenyl stannanuide; tetrabutylammonium | |
| difluoro(triphenyl)stannanuide;tetrabutylazanium |
Safety and Handling
TSCA : No