missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Difluorotriphenylsilicate 97.0+%, TCI America™

Catalog No. T19095G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19095G 5 g
T190925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T19095G Supplier TCI America Supplier No. T19095G
Only null left

Chemical Identifiers

CAS 163931-61-1
Molecular Formula C34H51F2NSi
Molecular Weight (g/mol) 539.871
MDL Number MFCD00274218
InChI Key RQBKGJOQACIQDG-UHFFFAOYSA-N
Synonym TBAT
PubChem CID 9893474
IUPAC Name difluoro(triphenyl)silanuide;tetrabutylazanium
SMILES CCCC[N+](CCCC)(CCCC)CCCC.C1=CC=C(C=C1)[Si-](C2=CC=CC=C2)(C3=CC=CC=C3)(F)F

Specifications

Melting Point 156°C
Color White
Quantity 5 g
UN Number 3261
Formula Weight 539.87
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Difluorotriphenylsilicate