missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium chloride hydrate, 96%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
Specifications
Specifications
| Melting Point | 37.0°C to 39.0°C |
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 95% min. (On dried substance) (Argentometry) |
| Packaging | Glass Bottle |
| Quantity | 100 g |
| Formula Weight | 295.94 |
| Percent Purity | 96% |
| Physical Form | Low Melting Solid |
| Chemical Name or Material | Tetrabutylammonium Chloride Hydrate |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for |
RUO – Research Use Only