missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium chloride, For ion pair chromatography, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111012467
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012467 10 g
111012468 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111012467 Supplier Merck Emd Millipore Supplier No. 8685210GF

Tetrabutylammonium chloride (TBACl) is a quaternary ammonium salt.g

Chemical Identifiers

MDL Number MFCD00011635

Specifications

CAS 1112-67-0
Flash Point 113°C - Closed Cup
Linear Formula [CH3(CH2)3]4 NCl
Quantity 10 g
Formula Weight 277.92
Percent Purity ≥99.0% (AT); ≥99.0%
Grade For ion pair chromatography