missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Bromide 98.0+%, TCI America™

Catalog No. T005425G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T005425G 25 g
T0054100G 100 g
T0054500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T005425G Supplier TCI America Supplier No. T005425G

Chemical Identifiers

CAS 1643-19-2
Molecular Formula C16H36BrN
Molecular Weight (g/mol) 322.375
MDL Number MFCD00011633
InChI Key JRMUNVKIHCOMHV-UHFFFAOYSA-M
Synonym tetrabutylammonium bromide, tetra-n-butylammonium bromide, tbab, tetrabutylazanium bromide, tetrabutyl ammonium bromide, tetrabutyl-ammonium bromide, aliquat 100, tbabr, ipc-tba-br, unii-vjz168i98r
PubChem CID 74236
ChEBI CHEBI:51993
IUPAC Name tetrabutylazanium;bromide
SMILES CCCC[N+](CCCC)(CCCC)CCCC.[Br-]

Specifications

Melting Point 103°C
Color White
pH 3.5 to 7.0 (50g/L H2O 25°C)
Quantity 25 g
Formula Weight 322.38
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Bromide
EINECSNumber (2)-0186
TSCA Yes