missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium bromide, For ion pair chromatography, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. p-7187766
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
50 g
Catalog No. Quantity
111013300 10 g
111013301 50 g
2 options

Catalog No. 111013300

Supplier: Merck Emd Millipore 8685710GF

Only null left

Chemical Identifiers

MDL Number MFCD00011633

Specifications

Melting Point 102°C to 106°C; 102°C to 106°C
CAS 1643-19-2
Linear Formula (CH3 CH2 CH2 CH2)4 N(Br)
Quantity 10 g
Formula Weight 322.37
Percent Purity ≥99.0% (AT); ≥99.0%
Grade For ion pair chromatography