missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Borohydride 96.0+%, TCI America™

Catalog No. T09175G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T09175G 5 g
T091725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T09175G Supplier TCI America Supplier No. T09175G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33725-74-5
Molecular Formula C16H40BN
Molecular Weight (g/mol) 257.31
MDL Number MFCD00012035
InChI Key GMBOFJFPOCGSOI-UHFFFAOYSA-N
Synonym tetrabutylammonium borohydride, tetra-n-butylammonium borohydride, n,n,n-tributylbutan-1-aminium tetrahydroborate, boron 1-; tetrabutylazanium, boron 1-; tetrabutylammonium
PubChem CID 9881569
IUPAC Name tetrabutylazanium boranuide
SMILES [BH4-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 126°C
Color White
Quantity 5 g
UN Number 3131
Formula Weight 257.31
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium Borohydride [Reducing Reagent]
EINECSNumber (2)-0186&(2)-3044
TSCA Yes