missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium Bis(maleonitriledithiolato)nickel(III) Complex 97.0+%, TCI America™
Supplier: TCI America T14151G
Specifications
| Tetrabutylammonium Bis(maleonitriledithiolato)nickel(III) Complex | |
| Black | |
| MFCD00191492 | |
| JETKTJCVEPVWKF-UHFFFAOYSA-J | |
| 1,2-dicyanoethene-1,2-dithiolate;nickel(3+);tetrabutylazanium | |
| 132769570 | |
| ≥97.0% (T) |
| 55401-12-2 | |
| C24H36N5NiS4 | |
| 1 g | |
| CCCC[N+](CCCC)(CCCC)CCCC.C(#N)C(=C(C#N)[S-])[S-].C(#N)C(=C(C#N)[S-])[S-].[Ni+3] | |
| 581.52 | |
| 581.52 | |
| Crystalline Powder |
Chemical Identifiers
| 55401-12-2 | |
| 581.52 | |
| JETKTJCVEPVWKF-UHFFFAOYSA-J | |
| 1,2-dicyanoethene-1,2-dithiolate;nickel(3+);tetrabutylazanium |
| C24H36N5NiS4 | |
| MFCD00191492 | |
| 132769570 | |
| CCCC[N+](CCCC)(CCCC)CCCC.C(#N)C(=C(C#N)[S-])[S-].C(#N)C(=C(C#N)[S-])[S-].[Ni+3] |
Safety and Handling
TSCA : No