missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Bis(3,4,6-trichloro-1,2-benzenedithiolato)nickelate 95.0+%, TCI America™

Catalog No. T3205200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3205200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3205200MG Supplier TCI America Supplier No. T3205200MG

Chemical Identifiers

CAS 87314-12-3
Molecular Formula C28H38Cl6NNiS4
Molecular Weight (g/mol) 788.252
InChI Key WIBIOFAUOGMVAE-UHFFFAOYSA-J
Synonym Tetrabutylammonium Bis(3,4,6-trichloro-1,2-benzenedithiolato)nickel(III) Complex
PubChem CID 91972100
IUPAC Name nickel(3+);tetrabutylazanium;3,4,6-trichlorobenzene-1,2-dithiolate
SMILES CCCC[N+](CCCC)(CCCC)CCCC.C1=C(C(=C(C(=C1Cl)Cl)[S-])[S-])Cl.C1=C(C(=C(C(=C1Cl)Cl)[S-])[S-])Cl.[Ni+3]

Specifications

Quantity 200 mg
Physical Form Crystal-Powder at 20°C
Formula Weight 788.25
Percent Purity ≥95.0% (T)
Chemical Name or Material Tetrabutylammonium Bis(3,4,6-trichloro-1,2-benzenedithiolato)nickelate