missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Bifluoride 95.0+%, TCI America™

Catalog No. T12955G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12955G 5 g
T129525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T12955G Supplier TCI America Supplier No. T12955G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23868-34-0
Molecular Formula C16H37F2N
Molecular Weight (g/mol) 281.476
MDL Number MFCD00077877
InChI Key ZHBDKVWQJKYIFF-UHFFFAOYSA-M
Synonym Tetrabutylammonium Hydrogen Difluoride
PubChem CID 10891295
IUPAC Name tetrabutylazanium;fluoride;hydrofluoride
SMILES CCCC[N+](CCCC)(CCCC)CCCC.F.[F-]

Specifications

Quantity 5 g
UN Number 1760
Formula Weight 281.48
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material Tetrabutylammonium Bifluoride
EINECSNumber (2)-0186
TSCA No