missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium Azide 95.0+%, TCI America™
$177.08 - $626.32
Chemical Identifiers
| CAS | 993-22-6 |
|---|---|
| Molecular Formula | C16H36N4 |
| Molecular Weight (g/mol) | 284.492 |
| MDL Number | MFCD00060069 |
| InChI Key | GMRIOAVKKGNMMV-UHFFFAOYSA-N |
| PubChem CID | 5743232 |
| IUPAC Name | tetrabutylazanium;azide |
| SMILES | CCCC[N+](CCCC)(CCCC)CCCC.[N-]=[N+]=[N-] |
Chemical Identifiers
| 993-22-6 | |
| 284.492 | |
| GMRIOAVKKGNMMV-UHFFFAOYSA-N | |
| tetrabutylazanium;azide |
| C16H36N4 | |
| MFCD00060069 | |
| 5743232 | |
| CCCC[N+](CCCC)(CCCC)CCCC.[N-]=[N+]=[N-] |
Specifications
| 993-22-6 | |
| White-Yellow | |
| MFCD00060069 | |
| 2811 | |
| CCCC[N+](CCCC)(CCCC)CCCC.[N-]=[N+]=[N-] | |
| 284.492 | |
| 284.49 | |
| Crystalline Powder |
| 85°C | |
| C16H36N4 | |
| 5 g | |
| GMRIOAVKKGNMMV-UHFFFAOYSA-N | |
| tetrabutylazanium;azide | |
| 5743232 | |
| ≥95.0% (T) | |
| Tetrabutylammonium Azide |
Safety and Handling
EINECSNumber : (2)-0186
TSCA : Yes
Recommended Storage : Refrigerator