missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium Acetate 90.0+%, TCI America™
$221.83 - $641.24
Chemical Identifiers
| CAS | 10534-59-5 |
|---|---|
| Molecular Formula | C18H39NO2 |
| Molecular Weight (g/mol) | 301.52 |
| MDL Number | MFCD00043208 |
| InChI Key | MCZDHTKJGDCTAE-UHFFFAOYSA-M |
| Synonym | tetrabutylammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate, tetra-n-butylammonium acetate, tetrabutylazanium acetate, tetrabutyl ammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate 1:1, tetrabutylammonium ion acetate, 2-phosphonooxy acrylic acid cyclohexylamine salt 1:1, n,n,n-tributylbutan-1-aminiumacetate, tetrabutylazanium ethanoate |
| PubChem CID | 82707 |
| IUPAC Name | tetrabutylazanium acetate |
| SMILES | CC([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC |
Chemical Identifiers
| 10534-59-5 | |
| 301.52 | |
| MCZDHTKJGDCTAE-UHFFFAOYSA-M | |
| 82707 | |
| CC([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC |
| C18H39NO2 | |
| MFCD00043208 | |
| tetrabutylammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate, tetra-n-butylammonium acetate, tetrabutylazanium acetate, tetrabutyl ammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate 1:1, tetrabutylammonium ion acetate, 2-phosphonooxy acrylic acid cyclohexylamine salt 1:1, n,n,n-tributylbutan-1-aminiumacetate, tetrabutylazanium ethanoate | |
| tetrabutylazanium acetate |
Specifications
| 10534-59-5 | |
| White-Yellow | |
| MFCD00043208 | |
| tetrabutylammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate, tetra-n-butylammonium acetate, tetrabutylazanium acetate, tetrabutyl ammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate 1:1, tetrabutylammonium ion acetate, 2-phosphonooxy acrylic acid cyclohexylamine salt 1:1, n,n,n-tributylbutan-1-aminiumacetate, tetrabutylazanium ethanoate | |
| CC([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC | |
| 301.52 | |
| 301.52 | |
| Paste |
| 98°C | |
| C18H39NO2 | |
| 25 g | |
| MCZDHTKJGDCTAE-UHFFFAOYSA-M | |
| tetrabutylazanium acetate | |
| 82707 | |
| ≥90.0% (T) | |
| Tetrabutylammonium Acetate |
Safety and Handling
EINECSNumber : (2)-0186
TSCA : Yes