missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutyl Orthotitanate, TCI America™

Catalog No. p-7118652
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B074225G 25 g
B0742500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B074225G Supplier TCI America Supplier No. B074225G
Only null left

Chemical Identifiers

CAS 5593-70-4
Molecular Formula C16H40O4Ti
Molecular Weight (g/mol) 344.359
MDL Number MFCD00009433
InChI Key FPCJKVGGYOAWIZ-UHFFFAOYSA-N
Synonym butyl titanate, butan-1-ol; titanium, titanium n-butoxide, tetrabutyl orthotitanate, tetrabutyl titanate, 1-butanol; titanium, tetra butyl orthotitanate, tetrabutyl ortho-titanate
PubChem CID 25203927
IUPAC Name butan-1-ol;titanium
SMILES CCCCO.CCCCO.CCCCO.CCCCO.[Ti]

Specifications

Melting Point -40°C
Color Yellow
Boiling Point 185°C
Quantity 25 g
UN Number 1993
Formula Weight 340.33
Physical Form Liquid
Chemical Name or Material Tetrabutyl Orthotitanate
EINECSNumber (2)-0228&(2)-2150
RTECSNumber XR1585000
TSCA Yes