missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutyl Orthosilicate 98.0+%, TCI America™

Catalog No. T112625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T112625ML 25 mL
T1126500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T112625ML Supplier TCI America Supplier No. T112625ML

Chemical Identifiers

CAS 4766-57-8
Molecular Formula C16H36O4Si
Molecular Weight (g/mol) 320.545
MDL Number MFCD00009432
InChI Key UQMOLLPKNHFRAC-UHFFFAOYSA-N
Synonym tetrabutyl orthosilicate, tetrabutoxysilane, tetra-n-butoxysilane, silane, tetrabutoxy, silicon tetrabutoxide, butyl silicate buo 4si, silicic acid h4sio4 , tetrabutyl ester, butyl silicate c4h9o 4si, butyl silicate, tetrabutyl orthosilice
PubChem CID 78500
IUPAC Name tetrabutyl silicate
SMILES CCCCO[Si](OCCCC)(OCCCC)OCCCC

Specifications

Color Colorless
Boiling Point 277°C
Quantity 25 mL
UN Number 1760
Formula Weight 320.55
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetrabutyl Orthosilicate
EINECSNumber (2)-2048
TSCA Yes