missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabromophenol Blue 95.0+%, TCI America™
$359.76 - $359.76
Chemical Identifiers
| CAS | 4430-25-5 |
|---|---|
| Molecular Formula | C19H6Br8O5S |
| Molecular Weight (g/mol) | 985.544 |
| MDL Number | MFCD00005876 |
| InChI Key | QPMIVFWZGPTDPN-UHFFFAOYSA-N |
| Synonym | Tetrabromophenoltetrabromosulfophthalein, TBPB |
| PubChem CID | 78159 |
| IUPAC Name | 2,6-dibromo-4-[4,5,6,7-tetrabromo-3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol |
| SMILES | C1=C(C=C(C(=C1Br)O)Br)C2(C3=C(C(=C(C(=C3Br)Br)Br)Br)S(=O)(=O)O2)C4=CC(=C(C(=C4)Br)O)Br |
Chemical Identifiers
| 4430-25-5 | |
| 985.544 | |
| QPMIVFWZGPTDPN-UHFFFAOYSA-N | |
| 78159 | |
| C1=C(C=C(C(=C1Br)O)Br)C2(C3=C(C(=C(C(=C3Br)Br)Br)Br)S(=O)(=O)O2)C4=CC(=C(C(=C4)Br)O)Br |
| C19H6Br8O5S | |
| MFCD00005876 | |
| Tetrabromophenoltetrabromosulfophthalein, TBPB | |
| 2,6-dibromo-4-[4,5,6,7-tetrabromo-3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol |
Specifications
| 204°C | |
| C19H6Br8O5S | |
| Tetrabromophenoltetrabromosulfophthalein, TBPB | |
| C1=C(C=C(C(=C1Br)O)Br)C2(C3=C(C(=C(C(=C3Br)Br)Br)Br)S(=O)(=O)O2)C4=CC(=C(C(=C4)Br)O)Br | |
| 985.544 | |
| 985.54 | |
| 5 g | |
| Tetrabromophenol Blue |
| 4430-25-5 | |
| MFCD00005876 | |
| QPMIVFWZGPTDPN-UHFFFAOYSA-N | |
| 2,6-dibromo-4-[4,5,6,7-tetrabromo-3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol | |
| 78159 | |
| ≥95.0% (HPLC) | |
| Crystal/Powder |
Safety and Handling
EINECSNumber : (4)-0906&(5)-3568
TSCA : Yes