missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Tetrabromophenol Blue, pure
Chemical Identifiers
| CAS | 4430-25-5 |
|---|---|
| Molecular Formula | C19H6Br8O5S |
| Molecular Weight (g/mol) | 985.53 |
| InChI Key | QPMIVFWZGPTDPN-UHFFFAOYSA-N |
| Synonym | tetrabromophenol blue, 4,5,6,7-tetrabromo-3,3-bis 3,5-dibromo-4-hydroxyphenyl-3h-benzo c 1,2 oxathiole 1,1-dioxide, tetrabromphenol tetrabromsulfonphthalein, unii-s6fo6931n7, ccris 5488, 3',3,5',5-tetrabromophenol-4,5,6,7-tetrabromosulfonephthalein, phenol, 4,4'-4,5,6,7-tetrabromo-1,1-dioxido-3h-2,1-benzoxathiol-3-ylidene bis 2,6-dibromo, 4,4'-4,5,6,7-tetrabromo-3h-2,1-benzoxathiol-3-ylidene bis 2,6-dibromophenol s,s-dioxide |
| PubChem CID | 78159 |
| IUPAC Name | 2,6-dibromo-4-[4,5,6,7-tetrabromo-3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol |
| SMILES | C1=C(C=C(C(=C1Br)O)Br)C2(C3=C(C(=C(C(=C3Br)Br)Br)Br)S(=O)(=O)O2)C4=CC(=C(C(=C4)Br)O)Br |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC189910050
|
Thermo Scientific™
189910050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC189910250
|
Thermo Scientific™
189910250 |
25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 4430-25-5 | |
| 985.53 | |
| tetrabromophenol blue, 4,5,6,7-tetrabromo-3,3-bis 3,5-dibromo-4-hydroxyphenyl-3h-benzo c 1,2 oxathiole 1,1-dioxide, tetrabromphenol tetrabromsulfonphthalein, unii-s6fo6931n7, ccris 5488, 3',3,5',5-tetrabromophenol-4,5,6,7-tetrabromosulfonephthalein, phenol, 4,4'-4,5,6,7-tetrabromo-1,1-dioxido-3h-2,1-benzoxathiol-3-ylidene bis 2,6-dibromo, 4,4'-4,5,6,7-tetrabromo-3h-2,1-benzoxathiol-3-ylidene bis 2,6-dibromophenol s,s-dioxide | |
| 2,6-dibromo-4-[4,5,6,7-tetrabromo-3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol |
| C19H6Br8O5S | |
| QPMIVFWZGPTDPN-UHFFFAOYSA-N | |
| 78159 | |
| C1=C(C=C(C(=C1Br)O)Br)C2(C3=C(C(=C(C(=C3Br)Br)Br)Br)S(=O)(=O)O2)C4=CC(=C(C(=C4)Br)O)Br |
Specifications
| 4430-25-5 | |
| Glass bottle | |
| 5g | |
| tetrabromophenol blue, 4,5,6,7-tetrabromo-3,3-bis 3,5-dibromo-4-hydroxyphenyl-3h-benzo c 1,2 oxathiole 1,1-dioxide, tetrabromphenol tetrabromsulfonphthalein, unii-s6fo6931n7, ccris 5488, 3',3,5',5-tetrabromophenol-4,5,6,7-tetrabromosulfonephthalein, phenol, 4,4'-4,5,6,7-tetrabromo-1,1-dioxido-3h-2,1-benzoxathiol-3-ylidene bis 2,6-dibromo, 4,4'-4,5,6,7-tetrabromo-3h-2,1-benzoxathiol-3-ylidene bis 2,6-dibromophenol s,s-dioxide | |
| QPMIVFWZGPTDPN-UHFFFAOYSA-N | |
| 2,6-dibromo-4-[4,5,6,7-tetrabromo-3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol | |
| 78159 | |
| Pure |
| Authentic | |
| C19H6Br8O5S | |
| 19,II,107 | |
| Solubility in water: soluble in water. | |
| C1=C(C=C(C(=C1Br)O)Br)C2(C3=C(C(=C(C(=C3Br)Br)Br)Br)S(=O)(=O)O2)C4=CC(=C(C(=C4)Br)O)Br | |
| 985.53 | |
| 985.53 | |
| Tetrabromophenol Blue |
Safety and Handling
EINECSNumber : 224-622-9
RUO â Research Use Only