missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tetrabromophenol Blue, pure

Catalog No. AC189910050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC189910050 5 g Glass bottle
AC189910250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC189910050 Supplier Thermo Scientific Chemicals Supplier No. 189910050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4430-25-5 | C19H6Br8O5S | 985.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4430-25-5
Molecular Formula C19H6Br8O5S
Molecular Weight (g/mol) 985.53
InChI Key QPMIVFWZGPTDPN-UHFFFAOYSA-N
Synonym tetrabromophenol blue, 4,5,6,7-tetrabromo-3,3-bis 3,5-dibromo-4-hydroxyphenyl-3h-benzo c 1,2 oxathiole 1,1-dioxide, tetrabromphenol tetrabromsulfonphthalein, unii-s6fo6931n7, ccris 5488, 3',3,5',5-tetrabromophenol-4,5,6,7-tetrabromosulfonephthalein, phenol, 4,4'-4,5,6,7-tetrabromo-1,1-dioxido-3h-2,1-benzoxathiol-3-ylidene bis 2,6-dibromo, 4,4'-4,5,6,7-tetrabromo-3h-2,1-benzoxathiol-3-ylidene bis 2,6-dibromophenol s,s-dioxide
PubChem CID 78159
IUPAC Name 2,6-dibromo-4-[4,5,6,7-tetrabromo-3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES C1=C(C=C(C(=C1Br)O)Br)C2(C3=C(C(=C(C(=C3Br)Br)Br)Br)S(=O)(=O)O2)C4=CC(=C(C(=C4)Br)O)Br

Specifications

Melting Point 203.0°C to 205.0°C
Color Beige
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5 g
Beilstein 19,II,107
Solubility Information Solubility in water: soluble in water.
Formula Weight 985.53
Grade Pure
Physical Form Powder
Chemical Name or Material Tetrabromophenol Blue
Show More Show Less
EINECSNumber 224-622-9

RUO – Research Use Only