missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tetrabromophenol Blue

CAS: 4430-25-5 | C19H6Br8O5S | 985.544 g/mol

$54.56 - $609.03

Chemical Identifiers

CAS 4430-25-5
Molecular Formula C19H6Br8O5S
Molecular Weight (g/mol) 985.544
MDL Number MFCD00005876
InChI Key QPMIVFWZGPTDPN-UHFFFAOYSA-N
Synonym 3,4,5,6,3',5',3'',5''-Octabromophenolsulfonphthalein
PubChem CID 78159
IUPAC Name 2,6-dibromo-4-[4,5,6,7-tetrabromo-3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES C1=C(C=C(C(=C1Br)O)Br)C2(C3=C(C(=C(C(=C3Br)Br)Br)Br)S(=O)(=O)O2)C4=CC(=C(C(=C4)Br)O)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2012303
View Documents
Thermo Scientific Chemicals
B2012303
1 g
Each for $54.56
Only null left
 
AAB2012306
View Documents
Thermo Scientific Chemicals
B2012306
5 g
Each for $161.52
Only null left
 
AAB2012314
View Documents
Thermo Scientific Chemicals
B2012314
25 g
Each for $609.03
Only null left
 
Description

Description

Tetrabromophenol Blue is used as acid-base indicator. It is used in oxidative- as well as in non-oxidative hair dye formulations. It is used as an industrial dye, a laboratory indicator, and a biological stain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrabromophenol Blue is used as acid-base indicator. It is used in oxidative- as well as in non-oxidative hair dye formulations. It is used as an industrial dye, a laboratory indicator, and a biological stain.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
Specifications

Chemical Identifiers

4430-25-5
985.544
QPMIVFWZGPTDPN-UHFFFAOYSA-N
78159
C1=C(C=C(C(=C1Br)O)Br)C2(C3=C(C(=C(C(=C3Br)Br)Br)Br)S(=O)(=O)O2)C4=CC(=C(C(=C4)Br)O)Br
C19H6Br8O5S
MFCD00005876
3,4,5,6,3',5',3'',5''-Octabromophenolsulfonphthalein
2,6-dibromo-4-[4,5,6,7-tetrabromo-3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol

Specifications

203°C to 205°C (decomposition)
C19H6Br8O5S
378438
Soluble in water.
C1=C(C=C(C(=C1Br)O)Br)C2(C3=C(C(=C(C(=C3Br)Br)Br)Br)S(=O)(=O)O2)C4=CC(=C(C(=C4)Br)O)Br
985.544
985.55
Tetrabromophenol Blue
4430-25-5
MFCD00005876
3,4,5,6,3',5',3'',5''-Octabromophenolsulfonphthalein
QPMIVFWZGPTDPN-UHFFFAOYSA-N
2,6-dibromo-4-[4,5,6,7-tetrabromo-3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
78159
1 g
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 224-622-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only