missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tetrabromophenol Blue

Catalog No. AAB2012303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2012303 1 g
AAB2012306 5 g
AAB2012314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2012303 Supplier Thermo Scientific Chemicals Supplier No. B2012303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4430-25-5 | C19H6Br8O5S | 985.544 g/mol

Tetrabromophenol Blue is used as acid-base indicator. It is used in oxidative- as well as in non-oxidative hair dye formulations. It is used as an industrial dye, a laboratory indicator, and a biological stain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrabromophenol Blue is used as acid-base indicator. It is used in oxidative- as well as in non-oxidative hair dye formulations. It is used as an industrial dye, a laboratory indicator, and a biological stain.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.

Chemical Identifiers

CAS 4430-25-5
Molecular Formula C19H6Br8O5S
Molecular Weight (g/mol) 985.544
MDL Number MFCD00005876
InChI Key QPMIVFWZGPTDPN-UHFFFAOYSA-N
Synonym 3,4,5,6,3',5',3'',5''-Octabromophenolsulfonphthalein
PubChem CID 78159
IUPAC Name 2,6-dibromo-4-[4,5,6,7-tetrabromo-3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES C1=C(C=C(C(=C1Br)O)Br)C2(C3=C(C(=C(C(=C3Br)Br)Br)Br)S(=O)(=O)O2)C4=CC(=C(C(=C4)Br)O)Br

Specifications

Melting Point 203°C to 205°C (decomposition)
Beilstein 378438
Solubility Information Soluble in water.
Formula Weight 985.55
Quantity 1 g
Chemical Name or Material Tetrabromophenol Blue
EINECSNumber 224-622-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only