missing translation for 'onlineSavingsMsg'
Learn More

Tetraammineplatinum(II) nitrate, Premion™, 99.99% (metals basis), Pt 50% min

Catalog No. AA4389706
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4389706 5 g
AA4389701 0.25 g
AA4389703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4389706 Supplier Thermo Scientific Chemicals Supplier No. 04389706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20634-12-2 | H12N6O6Pt | 387.22 g/mol

Tetraammineplatinum(II) nitrate is widely utilized as a precursor to bimetallic Pt/Zn catalysts for the selective hydrogenation of crotonaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20634-12-2
Molecular Formula H12N6O6Pt
Molecular Weight (g/mol) 387.22
MDL Number MFCD00011624
InChI Key RBAKORNXYLGSJB-UHFFFAOYSA-N
Synonym tetraammineplatinum ii nitrate, tetraammineplatinum dinitrate, tetrammineplatinum ii nitrate, tetraammineplatinum ii nitrate 1g, tetraammineplatinum ii nitrate, premion, platinum 2+ tetraamine dinitrate, tetraammineplatinum ii nitrate solution, pt w/w cont. pt
PubChem CID 146543
SMILES N.N.N.N.[Pt++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 262°C (decomposition)
Color White to Yellow
Physical Form Crystals
Quantity 5 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in water.
IUPAC Name azane;platinum(2+);dinitrate
Formula Weight 387.21
Chemical Name or Material Tetraammineplatinum(II) nitrate
Hazard Category H272-H315-H317-H319-H335
Precautionary Statement P210-P220-P221-P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P370+P378q-P501c
DOTInformation DOT Class: 5.1; Packaging Group: II
EINECSNumber 243-929-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only