missing translation for 'onlineSavingsMsg'
Learn More

Tetraammineplatinum(II) hydroxide hydrate, Pt 58% min

Catalog No. p-7080078
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4631903 1 g
AA4631906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4631903 Supplier Thermo Scientific Chemicals Supplier No. 04631903
Only null left

CAS: 312695-70-8 | H14N4O2Pt | 297.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 312695-70-8
Molecular Formula H14N4O2Pt
Molecular Weight (g/mol) 297.22
MDL Number MFCD00151253
InChI Key VSKCDODNDPOZKS-UHFFFAOYSA-L
IUPAC Name platinum(2+) tetraamine dihydroxide
SMILES N.N.N.N.[OH-].[OH-].[Pt++]

Specifications

Color White
Linear Formula (NH3)4Pt(OH)2·xH2O
Quantity 1 g
Formula Weight 297.23 (Anhydrous)
Concentration or Composition (by Analyte or Components) Pt 58% min
Physical Form Powder
Chemical Name or Material Tetraammineplatinum (II) hydroxide hydrate
Hazard Category H315-H317-H319-H335
Hazard Statement GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
EINECSNumber 253-823-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only