missing translation for 'onlineSavingsMsg'
Learn More

Tetraammineplatinum(II) chloride monohydrate

Catalog No. p-7048441
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1054403 1 g
AA1054406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1054403 Supplier Thermo Scientific Chemicals Supplier No. 01054403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13933-33-0 | Cl2H14N4OPt | 352.12 g/mol

Employed in the synthesis of cisplatin, and platinum included carbon - carbon materials with metal/metal oxide nanoparticles. Useful for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13933-33-0
Molecular Formula Cl2H14N4OPt
Molecular Weight (g/mol) 352.12
MDL Number MFCD00149947
InChI Key GWBDNMCYHWRSOH-UHFFFAOYSA-L
Synonym azane;platinum 2+ ;dichloride;hydrate, tetrammineplatinum ii chloride hydrate, tetraammineplatinum ii chloride monohydrate, platinum 2+ ion tetraamine hydrate dichloride, platinum 2+ tetraamine hydrate dichloride, platinum 2+ chloride-ammonia-water 1/2/4/1, tetraamineplatinium ii chloride
PubChem CID 21946612
IUPAC Name azane;platinum(2+);dichloride;hydrate
SMILES N.N.N.N.O.[Cl-].[Cl-].[Pt++]

Specifications

Melting Point 250°C (dec.)
Color White through Cream
Physical Form Crystalline
Quantity 1 g
Solubility Information Soluble in water,ethanol and chloroform.
Formula Weight 352.13 (334.11 Anhydrous)
Sensitivity Hygroscopic
Density 2.74g/mL
Chemical Name or Material Tetraammineplatinum(II) chloride monohydrate
Hazard Category H290-H315-H319
Hazard Statement GHS H Statement
H290-H315-H319
May be corrosive to metals.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P390
EINECSNumber 237-706-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only