Learn More
Tetraammineplatinum(II) chloride hydrate, 99.995%, (trace metal basis), Thermo Scientific™
Chemical Identifiers
| CAS | 108374-32-9 |
|---|---|
| Molecular Formula | Cl2H12N4Pt·xH2O |
| Molecular Weight (g/mol) | 334.11 |
| Synonym | platinum 4+ ion hydrate hydrochloride tetraazanide |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC437005000
|
Thermo Scientific Chemicals
437005000 |
500mg | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC437000050
|
Thermo Scientific Chemicals
437000050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
for ultra trace analysisChemical Identifiers
| 108374-32-9 | |
| Cl2H12N4Pt·xH2O | |
| 334.11 | |
| platinum 4+ ion hydrate hydrochloride tetraazanide |
Specifications
| 108374-32-9 | |
| Glass bottle | |
| 500mg | |
| 334.11 | |
| 99.995% | |
| Tetraammineplatinum(II) chloride hydrate |
| 99.995% | |
| Cl2H12N4Pt·xH2O | |
| platinum 4+ ion hydrate hydrochloride tetraazanide | |
| 334.11 | |
| Trace Metal Basis |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a
GHS Signal Word: Warning
RUO â Research Use Only