missing translation for 'onlineSavingsMsg'
Learn More

Tetraammineplatinum(II) chloride hydrate, 99.995%, (trace metal basis)

Catalog No. AC437005000
Change view
Click to view available options
Quantity:
500 mg
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC437005000 500 mg Glass bottle
AC437000050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC437005000 Supplier Thermo Scientific Chemicals Supplier No. 437005000

CAS: 108374-32-9 | Cl2H12N4Pt | 334.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis

Chemical Identifiers

CAS 108374-32-9
Molecular Formula Cl2H12N4Pt
Molecular Weight (g/mol) 334.11
MDL Number MFCD00149947
InChI Key KHCPSOMSJYAQSY-UHFFFAOYSA-L
Synonym platinum 4+ ion hydrate hydrochloride tetraazanide
IUPAC Name platinum(2+) tetraamine dichloride
SMILES N.N.N.N.[Cl-].[Cl-].[Pt++]

Specifications

Color White
Assay Percent Range 99.995%
Packaging Glass bottle
Quantity 500 mg
Formula Weight 334.11
Percent Purity 99.995%
Grade Trace Metal Basis
Physical Form Crystalline Powder
Chemical Name or Material Tetraammineplatinum(II) chloride hydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a

RUO – Research Use Only