missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraallyltin 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 70°C |
| Quantity | 1 g |
| UN Number | 2788 |
| Formula Weight | 283.00 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Tetraallyltin |
Safety and Handling
| TSCA | No |