missing translation for 'onlineSavingsMsg'
Learn More

Tetraallyltin 97.0+%, TCI America™

Catalog No. T20091G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20091G 1 g
T20095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T20091G Supplier TCI America Supplier No. T20091G

Chemical Identifiers

CAS 7393-43-3
Molecular Formula C12H20Sn
Molecular Weight (g/mol) 283.00
MDL Number MFCD00008637
InChI Key XJPKDRJZNZMJQM-UHFFFAOYSA-N
Synonym tetraallyltin, tetraallylstannane, tetrallylstannane, stannane, tetra-2-propenyl, stannane, tetraallyl, acmc-209osy, tetrakis prop-2-enyl stannane, stannane,tetra-2-propen-1-yl, tetrakis prop-2-en-1-yl stannane
PubChem CID 81878
IUPAC Name tetrakis(prop-2-en-1-yl)stannane
SMILES C=CC[Sn](CC=C)(CC=C)CC=C

Specifications

Color Yellow
Boiling Point 70°C
Quantity 1 g
UN Number 2788
Formula Weight 283.00
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetraallyltin