Learn More
Tetraallylsilane, 97%
Synthesis of dendrimers | CAS: 1112-66-9 | C12H20Si | 192.377 g/mol
$95.68 - $330.11
Chemical Identifiers
| CAS | 1112-66-9 |
|---|---|
| Molecular Formula | C12H20Si |
| Molecular Weight (g/mol) | 192.377 |
| MDL Number | MFCD00048161 |
| InChI Key | AKRQMTFHUVDMIL-UHFFFAOYSA-N |
| Synonym | tetraallylsilane, silane, tetra-2-propenyl, tetrakis prop-2-en-1-yl silane, tetrakis prop-2-enyl silane, silane, tetraallyl, tetraallyl silane, zlchem 610, acmc-20ajk5, tetra 2-propenyl silane, silane,tetra-2-propen-1-yl |
| PubChem CID | 565641 |
| IUPAC Name | tetrakis(prop-2-enyl)silane |
| SMILES | C=CC[Si](CC=C)(CC=C)CC=C |
Description
It is used in synthesis of dendrimers and as an intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1112-66-9 | |
| 192.377 | |
| AKRQMTFHUVDMIL-UHFFFAOYSA-N | |
| 565641 | |
| C=CC[Si](CC=C)(CC=C)CC=C |
| C12H20Si | |
| MFCD00048161 | |
| tetraallylsilane, silane, tetra-2-propenyl, tetrakis prop-2-en-1-yl silane, tetrakis prop-2-enyl silane, silane, tetraallyl, tetraallyl silane, zlchem 610, acmc-20ajk5, tetra 2-propenyl silane, silane,tetra-2-propen-1-yl | |
| tetrakis(prop-2-enyl)silane |
Specifications
| 91°C (10mmHg) | |
| Liquid | |
| C12H20Si | |
| 77°C (171°F) | |
| Insoluble in water. | |
| C=CC[Si](CC=C)(CC=C)CC=C | |
| 192.377 | |
| 192.38 | |
| 1.485 | |
| Tetraallylsilane |
| 1112-66-9 | |
| 1 g | |
| MFCD00048161 | |
| tetraallylsilane, silane, tetra-2-propenyl, tetrakis prop-2-en-1-yl silane, tetrakis prop-2-enyl silane, silane, tetraallyl, tetraallyl silane, zlchem 610, acmc-20ajk5, tetra 2-propenyl silane, silane,tetra-2-propen-1-yl | |
| AKRQMTFHUVDMIL-UHFFFAOYSA-N | |
| tetrakis(prop-2-enyl)silane | |
| 565641 | |
| 97% | |
| 0.8314 g/mL |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only