missing translation for 'onlineSavingsMsg'
Learn More

Tetraallylsilane, 97%

Catalog No. AA4318303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4318303 1 g
AA4318306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4318303 Supplier Thermo Scientific Chemicals Supplier No. 04318303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Synthesis of dendrimers | CAS: 1112-66-9 | C12H20Si | 192.377 g/mol

It is used in synthesis of dendrimers and as an intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1112-66-9
Molecular Formula C12H20Si
Molecular Weight (g/mol) 192.377
MDL Number MFCD00048161
InChI Key AKRQMTFHUVDMIL-UHFFFAOYSA-N
Synonym tetraallylsilane, silane, tetra-2-propenyl, tetrakis prop-2-en-1-yl silane, tetrakis prop-2-enyl silane, silane, tetraallyl, tetraallyl silane, zlchem 610, acmc-20ajk5, tetra 2-propenyl silane, silane,tetra-2-propen-1-yl
PubChem CID 565641
IUPAC Name tetrakis(prop-2-enyl)silane
SMILES C=CC[Si](CC=C)(CC=C)CC=C

Specifications

Boiling Point 91°C (10mmHg)
Physical Form Liquid
Quantity 1 g
Flash Point 77°C (171°F)
Solubility Information Insoluble in water.
Formula Weight 192.38
Percent Purity 97%
Refractive Index 1.485
Density 0.8314 g/mL
Chemical Name or Material Tetraallylsilane
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only