missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetra-O-acetyl-beta-D-ribofuranose 98.0+%, TCI America™
$63.38 - $216.24
Chemical Identifiers
| CAS | 13035-61-5 |
|---|---|
| Molecular Formula | C13H18O9 |
| Molecular Weight (g/mol) | 318.278 |
| MDL Number | MFCD00005358 |
| InChI Key | IHNHAHWGVLXCCI-FDYHWXHSSA-N |
| Synonym | beta-d-ribofuranose 1,2,3,5-tetraacetate, tetra-o-acetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-beta-d-ribofuranose, tetraacetylribose, 1,2,3,5-tetraacetyl-.-d-ribofuranose, tetra-o-acetyl-b-d-ribofuranose, 1,2,3,5-tetraacetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-b-d-ribofuranose, .beta.-d-ribofuranose, tetraacetate, ribofuranose, tetraacetate, .beta.-d |
| PubChem CID | 83064 |
| IUPAC Name | [(2R,3R,4R,5S)-3,4,5-triacetyloxyoxolan-2-yl]methyl acetate |
| SMILES | CC(=O)OCC1C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C |
Chemical Identifiers
| 13035-61-5 | |
| 318.278 | |
| IHNHAHWGVLXCCI-FDYHWXHSSA-N | |
| 83064 | |
| CC(=O)OCC1C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C |
| C13H18O9 | |
| MFCD00005358 | |
| beta-d-ribofuranose 1,2,3,5-tetraacetate, tetra-o-acetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-beta-d-ribofuranose, tetraacetylribose, 1,2,3,5-tetraacetyl-.-d-ribofuranose, tetra-o-acetyl-b-d-ribofuranose, 1,2,3,5-tetraacetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-b-d-ribofuranose, .beta.-d-ribofuranose, tetraacetate, ribofuranose, tetraacetate, .beta.-d | |
| [(2R,3R,4R,5S)-3,4,5-triacetyloxyoxolan-2-yl]methyl acetate |
Specifications
| 13035-61-5 | |
| White | |
| MFCD00005358 | |
| beta-d-ribofuranose 1,2,3,5-tetraacetate, tetra-o-acetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-beta-d-ribofuranose, tetraacetylribose, 1,2,3,5-tetraacetyl-.-d-ribofuranose, tetra-o-acetyl-b-d-ribofuranose, 1,2,3,5-tetraacetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-b-d-ribofuranose, .beta.-d-ribofuranose, tetraacetate, ribofuranose, tetraacetate, .beta.-d | |
| CC(=O)OCC1C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C | |
| 318.278 | |
| 318.28 | |
| Crystalline Powder |
| 83°C | |
| C13H18O9 | |
| 5 g | |
| IHNHAHWGVLXCCI-FDYHWXHSSA-N | |
| [(2R,3R,4R,5S)-3,4,5-triacetyloxyoxolan-2-yl]methyl acetate | |
| 83064 | |
| ≥98.0% (GC) | |
| Tetra-O-acetyl-beta-D-ribofuranose |
Safety and Handling
TSCA : No