missing translation for 'onlineSavingsMsg'
Learn More

Tetra-O-acetyl-beta-D-ribofuranose 98.0+%, TCI America™

Catalog No. R00665G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R00665G 5 g
R006625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R00665G Supplier TCI America Supplier No. R00665G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13035-61-5 | C13H18O9 | 318.278 g/mol

Chemical Identifiers

CAS 13035-61-5
Molecular Formula C13H18O9
Molecular Weight (g/mol) 318.278
MDL Number MFCD00005358
InChI Key IHNHAHWGVLXCCI-FDYHWXHSSA-N
Synonym beta-d-ribofuranose 1,2,3,5-tetraacetate, tetra-o-acetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-beta-d-ribofuranose, tetraacetylribose, 1,2,3,5-tetraacetyl-.-d-ribofuranose, tetra-o-acetyl-b-d-ribofuranose, 1,2,3,5-tetraacetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-b-d-ribofuranose, .beta.-d-ribofuranose, tetraacetate, ribofuranose, tetraacetate, .beta.-d
PubChem CID 83064
IUPAC Name [(2R,3R,4R,5S)-3,4,5-triacetyloxyoxolan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 83°C
Color White
Quantity 5 g
Formula Weight 318.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetra-O-acetyl-beta-D-ribofuranose