missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-propylammonium hydroxide, 40% w/w aq. soln.

Catalog No. p-7049652
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1745614 25 g
AA1745622 100 g
AA1745636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1745614 Supplier Thermo Scientific Chemicals Supplier No. 01745614
Only null left

CAS: 4499-86-9 | C12H29NO | 203.37 g/mol

Tetra-n-propylammonium hydroxide is used as an intermediate, ion exchange agent, surface-active agent and solvent. Further, it is used as an antistatic agent, detergent sanitizers, softener for textiles and paper products, emulsifying agents and pigment dispersers. It finds application as curing accelerators. In addition to this, it acts as a phase transfer catalyst in organic synthesis to enhance the reaction rate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4499-86-9
Molecular Formula C12H29NO
Molecular Weight (g/mol) 203.37
MDL Number MFCD00009360
InChI Key LPSKDVINWQNWFE-UHFFFAOYSA-M
Synonym tetrapropylammonium hydroxide, tetra-n-propylammonium hydroxide, 1-propanaminium, n,n,n-tripropyl-, hydroxide, tpaoh, tetrapropylammonium oxide, unii-t686luy7nk, tetrapropylazanium hydroxide, t686luy7nk, ammonium, tetrapropyl-, hydroxide, n,n,n-tripropyl-1-propanaminium hydroxide
PubChem CID 20586
IUPAC Name tetrapropylazanium;hydroxide
SMILES [OH-].CCC[N+](CCC)(CCC)CCC

Specifications

Color Colorless to Yellow
Density 0.99 g/mL
Boiling Point 102°C
Refractive Index 1.3716
Linear Formula (CH3CH2CH2)4NOH
Quantity 25 g
UN Number UN3267
Beilstein 4207983
Sensitivity Air Sensitive
Solubility Information Miscible with strong electrolytes.
Formula Weight 203.37
Concentration or Composition (by Analyte or Components) 40% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Tetra-n-propylammonium hydroxide
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314
Causes severe skin burns and eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
EINECSNumber 224-800-6
RTECSNumber BS8400000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only