missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetra-n-propylammonium hydroxide, 1M aq. soln.
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 1 |
| Quantity | 25 g |
| UN Number | UN3267 |
| Beilstein | 4207983 |
| Sensitivity | Air Sensitive |
| Formula Weight | 203.37 |
| Concentration or Composition (by Analyte or Components) | 1M aq. soln. |
| Chemical Name or Material | Tetra-n-propylammonium hydroxide |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314 Causes severe skin burns and eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S. |
| EINECSNumber | 224-800-6 |
| RTECSNumber | BS8400000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only