missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-hexylammonium perchlorate

Catalog No. p-7049897
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3083006 5 g
AA3083014 25 g
AA3083022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3083006 Supplier Thermo Scientific Chemicals Supplier No. 03083006
Only null left

CAS: 4656-81-9 | C24H52ClNO4 | 454.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4656-81-9
Molecular Formula C24H52ClNO4
Molecular Weight (g/mol) 454.13
MDL Number MFCD00043185
InChI Key RLCKPGXHGKSGOS-UHFFFAOYSA-M
Synonym tetra-n-hexylammonium perchlorate, tetrahexylammonium perchlorate, tetrahexylazanium perchlorate, 1-hexanaminium, n,n,n-trihexyl-, perchlorate, n,n,n-trihexylhexan-1-aminiumperchlorate, 1-hexanaminium, n,n,n-trihexyl-, perchlorate 1:1, tetra-n-hexyl ammonium perchlorate
PubChem CID 78388
SMILES [O-][Cl](=O)(=O)=O.CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC

Specifications

Quantity 5 g
UN Number UN1479
IUPAC Name tetrahexylazanium;perchlorate
Formula Weight 454.13
Physical Form Crystalline
Chemical Name or Material Tetra-n-hexylammonium perchlorate
Hazard Category H272-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
EINECSNumber 225-095-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only