missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-hexylammonium iodide, 99%

CAS: 2138-24-1 | C24H52IN | 481.591 g/mol

Supplier:  Thermo Scientific Chemicals A1758914

 View more versions of this product

Catalog No. AAA1758914


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tetra-n-hexylammonium iodide
99°C to 105°C
MFCD00041981
Hygroscopic; Light sensitive
VRKHAMWCGMJAMI-UHFFFAOYSA-M
tetrahexylazanium;iodide
75056
99%
2138-24-1
C24H52IN
25 g
tetrahexylammonium iodide, tetra-n-hexylammonium iodide, 1-hexanaminium, n,n,n-trihexyl-, iodide, tetrahexylazanium iodide, 1-hexanaminium, n,n,n-trihexyl-, iodide 1:1, acmc-209fjk, tetrahexyl ammonium iodide, tetra-n-hexyl ammonium iodide, ammonium, tetrahexyl-, iodide, 1-hexanaminium,n,n-trihexyl-, iodide
CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC.[I-]
481.591
481.59

Chemical Identifiers

2138-24-1
481.591
VRKHAMWCGMJAMI-UHFFFAOYSA-M
75056
CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC.[I-]
C24H52IN
MFCD00041981
tetrahexylammonium iodide, tetra-n-hexylammonium iodide, 1-hexanaminium, n,n,n-trihexyl-, iodide, tetrahexylazanium iodide, 1-hexanaminium, n,n,n-trihexyl-, iodide 1:1, acmc-209fjk, tetrahexyl ammonium iodide, tetra-n-hexyl ammonium iodide, ammonium, tetrahexyl-, iodide, 1-hexanaminium,n,n-trihexyl-, iodide
tetrahexylazanium;iodide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 218-382-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only