missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-hexylammonium iodide, 99%

Catalog No. AAA1758906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1758906 5 g
AAA1758914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1758906 Supplier Thermo Scientific Chemicals Supplier No. A1758906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2138-24-1 | C24H52IN | 481.591 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2138-24-1
Molecular Formula C24H52IN
Molecular Weight (g/mol) 481.591
MDL Number MFCD00041981
InChI Key VRKHAMWCGMJAMI-UHFFFAOYSA-M
Synonym tetrahexylammonium iodide, tetra-n-hexylammonium iodide, 1-hexanaminium, n,n,n-trihexyl-, iodide, tetrahexylazanium iodide, 1-hexanaminium, n,n,n-trihexyl-, iodide 1:1, acmc-209fjk, tetrahexyl ammonium iodide, tetra-n-hexyl ammonium iodide, ammonium, tetrahexyl-, iodide, 1-hexanaminium,n,n-trihexyl-, iodide
PubChem CID 75056
IUPAC Name tetrahexylazanium;iodide
SMILES CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC.[I-]

Specifications

Melting Point 99°C to 105°C
Quantity 5 g
Sensitivity Hygroscopic; Light sensitive
Formula Weight 481.59
Percent Purity 99%
Chemical Name or Material Tetra-n-hexylammonium iodide
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 218-382-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only