missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-hexylammonium hydrogen sulfate, 98%

Catalog No. AA4388204
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4388204 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4388204 Supplier Thermo Scientific Chemicals Supplier No. 04388204

CAS: 32503-34-7 | C24H52NO4S | 450.74 g/mol

Tetra-n-hexylammonium hydrogen sulfate is used as a lipophilic phase-transfer catalyst. It is also used in coatings, printing ink, rubber, glass, leather and cosmetics. Further, it acts as an efficient catalyst in ruthenium-catalyzed reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32503-34-7
Molecular Formula C24H52NO4S
Molecular Weight (g/mol) 450.74
MDL Number MFCD00037675
InChI Key RULHPTADXJPDSN-UHFFFAOYSA-L
Synonym tetrahexylammonium hydrogensulfate, tetrahexylammonium hydrogen sulfate, tetra-n-hexylammonium hydrogen sulfate, tetrahexylammonium hydrogensulphate, 1-hexanaminium, n,n,n-trihexyl-, sulfate 1:1, tetrahexylammonium hydrogen sulphate, tetrahexylazanium sulfate, acmc-1ailg, tetra-n-hexyl-ammonium hydrogensulphate, tetra-n-hexylammonium hydrogen sulphate
PubChem CID 11015848
IUPAC Name hydrogen sulfate;tetrahexylazanium
SMILES [O-]S([O-])(=O)=O.CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC

Specifications

Melting Point 96°C to 102°C
Quantity 2 g
Solubility Information Slightly soluble in water.
Formula Weight 451.76
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Tetra-n-hexylammonium hydrogen sulfate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 251-069-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only