missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-hexylammonium bromide, 98%

CAS: 4328-13-6 | C24H52BrN | 434.59 g/mol

$108.21 - $1910.05

Chemical Identifiers

CAS 4328-13-6
Molecular Formula C24H52BrN
Molecular Weight (g/mol) 434.59
MDL Number MFCD00011858
InChI Key SYZCZDCAEVUSPM-UHFFFAOYSA-M
Synonym tetrahexylammonium bromide, tetrahexylazanium bromide, tetra-n-hexylammonium bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide 1:1, n,n,n-trihexylhexan-1-aminiumbromide, acmc-209jtn, tetra-n-hexylammonium bromide 10g, 1-hexanaminium,n,n,n-trihexyl-, bromide 1:1
PubChem CID 78026
IUPAC Name tetrahexylazanium;bromide
SMILES [Br-].CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1137209
View Documents
Thermo Scientific Chemicals
A1137209
10 g
Each for $108.21
Only null left
 
AAA1137218
View Documents
Thermo Scientific Chemicals
A1137218
50 g
Each for $384.84
Only null left
 
AAA1137230
View Documents
Thermo Scientific Chemicals
A1137230
250 g
Each for $1,910.05
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4328-13-6
434.59
SYZCZDCAEVUSPM-UHFFFAOYSA-M
78026
[Br-].CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC
C24H52BrN
MFCD00011858
tetrahexylammonium bromide, tetrahexylazanium bromide, tetra-n-hexylammonium bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide 1:1, n,n,n-trihexylhexan-1-aminiumbromide, acmc-209jtn, tetra-n-hexylammonium bromide 10g, 1-hexanaminium,n,n,n-trihexyl-, bromide 1:1
tetrahexylazanium;bromide

Specifications

4328-13-6
C24H52BrN
10 g
Hygroscopic
SYZCZDCAEVUSPM-UHFFFAOYSA-M
tetrahexylazanium;bromide
78026
98%
∼95°C to 100°C
MFCD00011858
3637302
tetrahexylammonium bromide, tetrahexylazanium bromide, tetra-n-hexylammonium bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide 1:1, n,n,n-trihexylhexan-1-aminiumbromide, acmc-209jtn, tetra-n-hexylammonium bromide 10g, 1-hexanaminium,n,n,n-trihexyl-, bromide 1:1
[Br-].CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC
434.59
434.6
Tetra-n-hexylammonium bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 224-363-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only