missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-hexylammonium bromide, 98%

Catalog No. AAA1137209
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1137209 10 g
AAA1137218 50 g
AAA1137230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1137209 Supplier Thermo Scientific Chemicals Supplier No. A1137209

CAS: 4328-13-6 | C24H52BrN | 434.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4328-13-6
Molecular Formula C24H52BrN
Molecular Weight (g/mol) 434.59
MDL Number MFCD00011858
InChI Key SYZCZDCAEVUSPM-UHFFFAOYSA-M
Synonym tetrahexylammonium bromide, tetrahexylazanium bromide, tetra-n-hexylammonium bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide, 1-hexanaminium, n,n,n-trihexyl-, bromide 1:1, n,n,n-trihexylhexan-1-aminiumbromide, acmc-209jtn, tetra-n-hexylammonium bromide 10g, 1-hexanaminium,n,n,n-trihexyl-, bromide 1:1
PubChem CID 78026
IUPAC Name tetrahexylazanium;bromide
SMILES [Br-].CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC

Specifications

Melting Point ∼95°C to 100°C
Quantity 10 g
Beilstein 3637302
Sensitivity Hygroscopic
Formula Weight 434.6
Percent Purity 98%
Chemical Name or Material Tetra-n-hexylammonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 224-363-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only