missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-dodecylammonium iodide, 98%

Catalog No. AAL0815304
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0815304 2 g
AAL0815309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0815304 Supplier Thermo Scientific Chemicals Supplier No. L0815304
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 40797-39-5 | C48H100IN | 818.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40797-39-5
Molecular Formula C48H100IN
Molecular Weight (g/mol) 818.24
MDL Number MFCD00051856
InChI Key OBMLOTDECOTAGU-UHFFFAOYSA-M
Synonym tetradodecylazanium iodide, tetra-n-dodecylammonium iodide, tetradodecylammonium iodide, tetradodecylazanium,iodide
PubChem CID 3084801
SMILES [I-].CCCCCCCCCCCC[N+](CCCCCCCCCCCC)(CCCCCCCCCCCC)CCCCCCCCCCCC

Specifications

Melting Point 114°C to 118°C
Linear Formula [CH3(CH2)11]4NI
Quantity 2 g
Beilstein 5844188
Sensitivity Light sensitive
IUPAC Name tetradodecylazanium iodide
Formula Weight 818.23
Percent Purity 98%
Chemical Name or Material Tetra-n-dodecylammonium iodide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 255-086-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only