missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylgermanium, 99%

Catalog No. AA3411906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3411906 5 g
AA3411914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3411906 Supplier Thermo Scientific Chemicals Supplier No. 03411906

CAS: 1067-42-1 | C16H36Ge | 301.094 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1067-42-1
Molecular Formula C16H36Ge
Molecular Weight (g/mol) 301.094
MDL Number MFCD00015224
InChI Key HDVLQIDIYKIVRE-UHFFFAOYSA-N
Synonym tetrabutylgermanium, germane, tetrabutyl, tetra-n-butylgermane, tetra-n-butylgermanium, tetrabutyl-german, tetrabuthylgermanium, tetrabutyl germanium
PubChem CID 70599
IUPAC Name tetrabutylgermane
SMILES CCCC[Ge](CCCC)(CCCC)CCCC

Specifications

Density 0.934 g/mL at 20°C
Boiling Point 277°C to 279°C
Flash Point >110°C (230°F)
Refractive Index 1.4571
Quantity 5 g
Formula Weight 301.05
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Tetra-n-butylgermanium
Hazard Category H302
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 213-929-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only