missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium trifluoromethanesulfonate, 99%

Catalog No. AAA1101306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1101306 5 g
AAA1101314 25 g
AAA1101322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1101306 Supplier Thermo Scientific Chemicals Supplier No. A1101306

CAS: 35895-70-6 | C18H36F3NO2 | 355.49 g/mol

Used as a catalyst for the reactions of condensation of alcohols and carboxylic acids, reaction of aromatic compounds with sulfonyl chlorides, cracking of alkanes, alkylation of alkenes, isomerisation of alkanes and trans-alkylation of aromatics, trans-bromination and other Friedel-Crafts reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a catalyst for the reactions of condensation of alcohols and carboxylic acids, reaction of aromatic compounds with sulfonyl chlorides, cracking of alkanes, alkylation of alkenes, isomerisation of alkanes and trans-alkylation of aromatics, trans-bromination and other Friedel-Crafts reactions.

Solubility
Miscible in watetr. Soluble in methanol(0.1 g/mL).

Notes
Hygroscopic. Keep container tightly closed in a cool, dry and well-ventilated place. Store away from oxidizing agents, air and water.

Chemical Identifiers

CAS 35895-70-6
Molecular Formula C18H36F3NO2
Molecular Weight (g/mol) 355.49
MDL Number MFCD00042585
InChI Key WTEXQPWIUJQYJQ-UHFFFAOYSA-M
Synonym tetrabutylammonium triflate, tetrabutylammonium trifluoromethanesulfonate, tetra-n-butylammonium trifluoromethanesulfonate, tetrabutylammonium ion triflate, trifluoromethanesulfonic acid tetrabutylammonium salt, acmc-1aglb, tetrakis but-1-yl ammonium triflate, tetrabutylazanium; trifluoromethanesulfonate, tetra-n-butylammonium trifluoromethanesulphonate
PubChem CID 2733306
IUPAC Name tetrabutylazanium;trifluoromethanesulfonate
SMILES [O-]C(=O)C(F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 112°C to 115°C
Quantity 5 g
Beilstein 4172978
Sensitivity Hygroscopic
Solubility Information Miscible in watetr. Soluble in methanol(0.1g/mL).
Formula Weight 391.54
Percent Purity 99%
Chemical Name or Material Tetra-n-butylammonium trifluoromethanesulfonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 252-783-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only