missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium sulfate, 50% w/w aq. soln.

Catalog No. AA41723AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41723AE 100 mL
AA41723AP 500 mL
AA41723K4 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA41723AE Supplier Thermo Scientific Chemicals Supplier No. 041723AE

Phase transfer catalyst, neutral pH | CAS: 2472-88-0 | C32H72N2O4S | 581.00 g/mol

Tetra-n-butylammonium sulfate is used as a phase transfer catalyst, neutral pH and organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2472-88-0
Molecular Formula C32H72N2O4S
Molecular Weight (g/mol) 581.00
MDL Number MFCD00216632
InChI Key ZXUCBXRTRRIBSO-UHFFFAOYSA-L
Synonym bis tetrabutylammonium sulphate, tetrabutylammonium sulfate, tetrabutylammonium sulfate solution, 1-butanaminium, n,n,n-tributyl-, sulfate 2:1, tetrabutyl ammoniumsulfat, tetrabutylammonium sulphate, acmc-1cdo3, tetrabutyl ammonium sulfate, tetrabutyl-ammonium sulfate, tetra-n-butylammonium sulfate
PubChem CID 11433187
IUPAC Name tetrabutylazanium;sulfate
SMILES [O-]S([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Density 1.01
Boiling Point 84°C
Odor Unpleasant
Refractive Index 1.417
Quantity 100 mL
Solubility Information Fully miscible.
Formula Weight 580.99
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material Tetra-n-butylammonium sulfate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 219-593-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only