missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium hexafluorophosphate, 98%

Catalog No. AAA1719636
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1719636 500 g
AAA1719614 25 g
AAA1719622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1719636 Supplier Thermo Scientific Chemicals Supplier No. A1719636
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3109-63-5 | C16H36F6NP | 387.44 g/mol

Tetra-n-butylammonium hexafluorophosphate is used as a supporting electrolyte in nonaqueous electrochemistry. It is chemically inert, thereby allowing the usage as an inert electrolyte with potential range. It enhances the performance of single-layer organic light emitting diodes (OLEDs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-butylammonium hexafluorophosphate is used as a supporting electrolyte in nonaqueous electrochemistry. It is chemically inert, thereby allowing the usage as an inert electrolyte with potential range. It enhances the performance of single-layer organic light emitting diodes (OLEDs).

Solubility
Soluble in acetone, acetonitrile and tetrhydrofuran.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 3109-63-5
Molecular Formula C16H36F6NP
Molecular Weight (g/mol) 387.44
MDL Number MFCD00011748
InChI Key BKBKEFQIOUYLBC-UHFFFAOYSA-N
Synonym tetrabutylammonium hexafluorophosphate, tetra-n-butylammonium hexafluorophosphate, tetrabutylammonium hexafluorophosphate v, 1-butanaminium, n,n,n-tributyl-, hexafluorophosphate 1-, 1-butanaminium, n,n,n-tributyl-, hexafluorophosphate 1-1:1, acmc-1ahjo, n,n,n-tributyl-1-butanaminium hexafluorophosphate, ksc225q6r, tetrabutylammoniumhexafluorophosphate, tetrabutylazanium hexafluorophosphate
PubChem CID 165075
SMILES F[P-](F)(F)(F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 242°C to 248°C
Linear Formula [CH3(CH2)3]4NPF6
Quantity 500 g
Sensitivity Hygroscopic
Solubility Information Soluble in acetone,acetonitrile and tetrhydrofuran.
IUPAC Name tetrabutylazanium;hexafluorophosphate
Formula Weight 387.44
Percent Purity 98%
Chemical Name or Material Tetra-n-butylammonium hexafluorophosphate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-472-6
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only