missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium dihydrogentrifluoride, tech. 90%, Thermo Scientific Chemicals

Catalog No. AAH2726606
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2726606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2726606 Supplier Thermo Scientific Chemicals Supplier No. H2726606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 99337-56-1 | Molecular Formula: C16H38F3N | Molecular Weight: 301.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99337-56-1
Molecular Formula C16H38F3N
Molecular Weight (g/mol) 301.48
MDL Number MFCD00145365
InChI Key MRXQMNWIADOAJY-UHFFFAOYSA-M
Synonym tetra-n-butylammonium dihydrogen trifluoride, tetra-n-butylammonium dihydrogentrifluoride, dihydrogen tetrabutylammonium fluoride, tetrabutylammonium dihydrogentrifluoride, tetrabutylammonium dihydrogen trifluoride, tetrabutylammonium fluoride dihydrofluoride, tetrabutylazanium fluoride dihydrofluoride, tetrabutylammoniumdihydrogentrifluoride, tetrabutylamonium dihydrogentrifluoride, tetrabutylammoniumdihydrogen trifluoride
PubChem CID 11748636
IUPAC Name tetrabutylazanium;fluoride;dihydrofluoride
SMILES F.F.[F-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Quantity 5 g
UN Number UN3265
Formula Weight 301.48
Percent Purity 90%
Grade Technical
Chemical Name or Material Tetra-n-butylammonium dihydrogentrifluoride
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only