missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetra-n-butylammonium dihydrogentrifluoride, 50-55% w/w soln. in 1,2-dichloroethane
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Flash Point | 13°C (55°F) |
| Quantity | 1 g |
| UN Number | UN2924 |
| IUPAC Name | tetrabutylazanium;fluoride;dihydrofluoride |
| Formula Weight | 301.48 |
| Concentration or Composition (by Analyte or Components) | 50 to 55% w/w soln. in 1,2-dichloroethane |
| Chemical Name or Material | Tetra-n-butylammonium dihydrogentrifluoride |
Safety and Handling
| Hazard Category | H224-H302+H312+H332-H314-H335-H350 |
| Hazard Statement | GHS H Statement H224-H301-H350-H314-H318 Extremely flammable liquid and vapor. Toxic if swallowed. May cause cancer. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only