missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium dihydrogentrifluoride, 50-55% w/w soln. in 1,2-dichloroethane

Catalog No. AAL1789106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1789106 5 g
AAL1789103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAL1789106 Supplier Thermo Scientific Chemicals Supplier No. L1789106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 99337-56-1 | C16H38F3N | 301.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99337-56-1
Molecular Formula C16H38F3N
Molecular Weight (g/mol) 301.48
MDL Number MFCD00145365
InChI Key MRXQMNWIADOAJY-UHFFFAOYSA-M
Synonym tetra-n-butylammonium dihydrogen trifluoride, tetra-n-butylammonium dihydrogentrifluoride, dihydrogen tetrabutylammonium fluoride, tetrabutylammonium dihydrogentrifluoride, tetrabutylammonium dihydrogen trifluoride, tetrabutylammonium fluoride dihydrofluoride, tetrabutylazanium fluoride dihydrofluoride, tetrabutylammoniumdihydrogentrifluoride, tetrabutylamonium dihydrogentrifluoride, tetrabutylammoniumdihydrogen trifluoride
PubChem CID 11748636
SMILES F.F.[F-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Flash Point 13°C (55°F)
Quantity 5 g
UN Number UN2924
IUPAC Name tetrabutylazanium dihydrofluoride fluoride
Formula Weight 301.48
Concentration or Composition (by Analyte or Components) 50 to 55% w/w soln. in 1,2-dichloroethane
Chemical Name or Material Tetra-n-butylammonium dihydrogentrifluoride
Hazard Category H224-H302+H312+H332-H314-H335-H350
Hazard Statement GHS H Statement
H224-H301-H350-H314-H318
Extremely flammable liquid and vapor.
Toxic if swallowed.
May cause cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only