missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium chloride hydrate

Catalog No. p-7049716
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1934706 5 g
AA1934714 25 g
AA1934722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1934706 Supplier Thermo Scientific Chemicals Supplier No. 01934706

CAS: 37451-68-6 | C16H36ClN | 277.92 g/mol

Tetra-n-butylammonium chloride hydrate is used with phosphorus pentoxide for greener deoxychlorination. It is used as a phase transfer catalyst for the preparation of 2-(3-pyridyl)-1-methylpyrrole by the reaction between 3-bromopyridine and 1-methyl-2pyrrolecarboxylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 37451-68-6
Molecular Formula C16H36ClN
Molecular Weight (g/mol) 277.92
MDL Number MFCD00011635
InChI Key NHGXDBSUJJNIRV-UHFFFAOYSA-M
Synonym tetrabutylammonium chloride hydrate, tetrabutyl ammonium chloride hydrate, tetrabutylammonium chloride monohydrate, tetra-n-butylammonium chloride hydrate, tetrabutylammoniumchlorideh2o, 1-butanaminium, n,n,n-tributyl-, chloride, monohydrate, tbac hydrate, bu4ncl.h2o, n-bu4ncl.h2o, ksc222g0b
PubChem CID 16212273
SMILES [Cl-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 73°C to 75°C
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in cold water.
IUPAC Name tetrabutylazanium chloride
Formula Weight 277.92 (Anhydrous)
Physical Form Crystalline
Chemical Name or Material Tetra-n-butylammonium chloride hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-195-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only